Tisk…
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 5 µm
Dodavatel: Avantor
EXL-1212-1002UEA 0 CZK
EXL-1212-1002U EXL-1212-1003U EXL-1212-1046U EXL-1212-1502U EXL-1212-1503U EXL-1212-1546U EXL-1212-2502U EXL-1212-2503U EXL-1212-2546U EXL-1212-0502U EXL-1212-0503U EXL-1212-0546U HICHACE12120102GD HICHACE12120103GD
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 5 µm
Kolony chromatografické
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS
More Product Information