Tisk…
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Dodavatel: Avantor
EXL-101-0202UEA 20660 CZK
EXL-101-0202U EXL-101-1502U EXL-101-1002U EXL-101-1003U EXL-101-1046U EXL-101-1203U EXL-101-1246U EXL-101-1503U EXL-101-1546U EXL-101-0302U EXL-101-0303U EXL-101-0346U EXL-101-3502U EXL-101-0502U EXL-101-0503U EXL-101-0546U EXL-101-7502U EXL-101-7503U EXL-101-7546U ACE-1P1-GD2U ACE-1P1-GD4U
VWR® ACE® Excel® C18 HPLC/UHPLC Columns, Analytical, 2 µm
Kolony chromatografické
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range ofMore Product Information