Tisk…

Anti-GOLGA4 / Golgin-245 Goat polyclonal antibody unconjugated

Dodavatel: Everest Biotech
EB06924
EVBIEB06924EA 9660 CZK
EVBIEB06924
Anti-GOLGA4 / Golgin-245 Goat polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: GOLGA4 / Golgin-245
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR