Tisk…
ATC probes, Eutech
Dodavatel: EUTECH
ECPH5TEM01P ECPHWPTEM01W
EUTEECPH5TEM01PEA 2010 CZK
EUTEECPH5TEM01P EUTEECPHWPTEM01W
ATC probes, Eutech
Elektrody Elektrody pH
ATC electrodes for use with Eutech pH 5+, pH 6+ and ION 6+ meters or for use with CyberScan pH 300 and 310 meters.