Tisk…
Conductivity meter, handheld, Eutech CyberScan COND 600
Dodavatel: EUTECH
ECCONWP60043K
EUTEECCONWP60043KEA 43820 CZK
EUTEECCONWP60043K
Conductivity meter, handheld, Eutech CyberScan COND 600
Meřiče elektrochemické Konduktometry
The waterproof, portable conductivity/TDS meter features a broad conductivity range with auto-ranging capabilities for more convenience.
  • Mega-memory space that stores/ recalls up to 500 data sets formatted to meet GLP standards
  • Wireless and cable-free data transfer with infrared IrDA technology.
  • Large, easy-to-read custom screen with simultaneous information display allows optimum viewing with user-friendly icons
  • HOLD function, auto-power off feature
  • Self-diagnostic with message codes for easy troubleshooting
  • Rugged and waterproof to IP67 standard with