Tisk…
Floor marking and striping rolls, DuraStripe® Supreme V
Dodavatel: ERGOMAT
DSV3100Y DSV2100O DSV2100G DSV2100B DSV2100BL DSV2100B/W DSV2100B/Y DSV2100G/W DSV2100R DSV2100R/W DSV2100W DSV2100Y DSV4100B DSV4100BL DSV4100G/W DSV4100R/W DSV4100B/W DSV4100B/Y DSV4100G DSV4100O DSV4100R DSV4100W DSV4100Y DSV34100Y DSV34100W DSV34100BLK/YE DSV3100O DSV3100R DSV3100RD/WH DSV3100W DSV34100B DSV34100BL DSV34100G DSV34100GRN/WH DSV34100O DSV34100R DSV34100RD/WH DSV34100B/W DSV3100G DSV3100GRN/WH DSV3100B DSV3100B/W DSV3100BL DSV3100BLK/YE
ERGODSV3100YEA 5310 CZK
ERGODSV3100Y ERGODSV2100O ERGODSV2100G ERGODSV2100B ERGODSV2100BL ERGODSV2100B/W ERGODSV2100B/Y ERGODSV2100G/W ERGODSV2100R ERGODSV2100R/W ERGODSV2100W ERGODSV2100Y ERGODSV4100B ERGODSV4100BL ERGODSV4100G/W ERGODSV4100R/W ERGODSV4100B/W ERGODSV4100B/Y ERGODSV4100G ERGODSV4100O ERGODSV4100R ERGODSV4100W ERGODSV4100Y ERGODSV34100Y ERGODSV34100W ERGODSV34100BLK/YE ERGODSV3100O ERGODSV3100R ERGODSV3100RD/WH ERGODSV3100W ERGODSV34100B ERGODSV34100BL ERGODSV34100G ERGODSV34100GRN/WH ERGODSV34100O ERGODSV34100R ERGODSV34100RD/WH ERGODSV34100B/W ERGODSV3100G ERGODSV3100GRN/WH ERGODSV3100B ERGODSV3100B/W ERGODSV3100BL ERGODSV3100BLK/YE
Floor marking and striping rolls, DuraStripe® Supreme V
Značení bezpečnostní
DuraStripe® is the easiest, fastest and least expensive floor marking system for industrial floors. The durability and vast size of the DuraStripe program makes it the only total floor marking solution available on the market today. Economic than painting the floors.
  • Ideal for less than perfect floors
  • Simple and easy peel-&-stick installation
  • No dry time, cure time, fumes or messy clean up to limit productivity
The Supreme V combines the adhesive from the original DuraStripe which is the superior choice under non ideal floor conditions along withMore Product Information