Tisk…
Restriction enzymes, Eco32I (EcoRV) (HC), Fermentas
Dodavatel: Thermo Fisher Scientific
ER0303
ER0303EA 6290 CZK
ER0303
Restriction enzymes, Eco32I (EcoRV) (HC), Fermentas
Činidla pro nukleové kyseliny Restriction Enzymes
Eco32I (EcoRV) restriction enzyme recognizes GAT^ATC sites and cuts best at 37 °C in R buffer Isoschizomers: EcoRV.
  • Superior quality—stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Used in molecular cloning, restriction site mapping, genotyping, southern blotting and restriction fragment length polymorphism (RFLP).
Objednat
Produktová specifikace
Environmentally Preferable
Aplikace
Název enzymu EcoRV
Zdroj
Overhang type Blunt
Kompatibilní buffer FastDigest Green Buffer or FastDigest Buffer (colorless)