Tisk…
Sample vials, economy processed
Dodavatel: EP SCIENTIFIC
SB36-0040 SB46-0040 TB36-0040
EPSCSB36-0040EA 15600 CZK
EPSCSB36-0040 EPSCSB46-0040 EPSCTB36-0040
Sample vials, economy processed
Vialky
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Open-top white PP cap with choice of standard 3,18 mm or thinner 1,52 mm PTFE-lined silicone septum
  • Directly compatible with automation; thin septa vial is optimised for easy needle puncture
  • Vial shoulder slope optimised to prevent air bubble entrapment
  • With unattached sample analysis labels for optional use
Processed to meet or exceed EPA performance based specifications for volatile organic analysis but withoutMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí NAR Certificate of Retirement