Tisk…
Snap vials, clean
Dodavatel: EP SCIENTIFIC
SS146-0040 SS236-0040 SS136-0040 SS246-0040 SS326-0020 SS336-0040 SS346-0040
EPSCSS146-0040EA 9040 CZK
EPSCSS146-0040 EPSCSS236-0040 EPSCSS136-0040 EPSCSS246-0040 EPSCSS326-0020 EPSCSS336-0040 EPSCSS346-0040
Snap vials, clean
Vialky
Clean snap VOA vials feature a snap-off septum guard that protects the septum from exposure to the general environment. The clean snap cap reduces the chance of volatile organic contaminants in the environment from passing through the septum and contaminating the sample during transport and storage. It also keeps the septum clean and free from foreign particles during handling. The clean snap cap easily peels away to provide access to the septum at the time of analysis.
  • Amber borosilicate glass vial is recommended for light sensitive samples
  • Septa
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí NAR Certificate of Retirement