Tisk…
Sampling bottles, narrow/wide neck, HDPE
Dodavatel: EP SCIENTIFIC
N411-0125 N411-0250 N319-1000 N311-0500 N311-0500BPC N311-1000 N311-1000BPC N319-0125 N311-0250 N319-0500 N311-0125BPC N411-0500 N319-0250 N311-0250BPC N301-0125 N301-0250 N301-0500 N301-1000 N311-0125
216-4433EA 11710 CZK
216-4433 216-4434 EPSCN319-1000 EPSCN311-0500 EPSCN311-0500BPC EPSCN311-1000 EPSCN311-1000BPC EPSCN319-0125 EPSCN311-0250 EPSCN319-0500 EPSCN311-0125BPC EPSCN411-0500 EPSCN319-0250 EPSCN311-0250BPC EPSCN301-0125 EPSCN301-0250 EPSCN301-0500 EPSCN301-1000 EPSCN311-0125
Sampling bottles, narrow/wide neck, HDPE
Láhve Láhve plastové
HDPE, with leak-proof PP/HDPE closure.
  • Narrow mouth for easy handling and precision pouring
  • Wide mouth for easy aseptic filling
  • Leakproof and break resistant
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Each container individually barcoded with lot number and unique container number
Recommended for water and other environmental liquid sample collections, especially for critical projects where sample containment and transport is of elevated concern.
Certifikace: Certificate of analysis (CofA) includedMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Certifikát ochrany životního prostředí IRECS; ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement