Tisk…
Sample container, oblong, with lined closure
Dodavatel: EP SCIENTIFIC
316-1000BPC 316-0125BPC 316-0250BPC 316-0500BPC
EPSC316-1000BPCEA 16240 CZK
EPSC316-1000BPC EPSC316-0125BPC EPSC316-0250BPC EPSC316-0500BPC
Sample container, oblong, with lined closure
Nádobky na vzorky Nádoby pro obecné použití
HDPE, natural, with LDPE-lined PP cap
  • Certified to meet EPA Performance Based Specifications for metals, cyanide and fluoride analyses
  • Caps assembled onto containers
  • Space-saving shape for optimal storage efficiency
These oblong wide-mouth bottles are excellent general purpose sample containers for liquid sampling for the purpose of inorganic analyte testing. The wide mouth feature makes this bottle ideal for sampling more viscous liquid samples like industrial wastes and sludges. Space-saving oblong shape and flat side panels allow bottles to beMore Product Information