Tisk…
Water sampling bottles, narrow neck, Cubitainer®
Dodavatel: EP SCIENTIFIC
314-0005 314-0001 314-0025 114-0001 114-0025 114-1000 314-1000
EPSC314-0005EA 3100 CZK
EPSC314-0005 EPSC314-0001 EPSC314-0025 EPSC114-0001 EPSC114-0025 EPSC114-1000 EPSC314-1000
Water sampling bottles, narrow neck, Cubitainer®
Láhve Láhve na odběr vzorků vody
Cubitainers™ are designed for the efficient storage and transport. Recommended for collecting water samples and other low viscosity liquids to be analysed for inorganic trace analytes.
  • Light weight and space-efficient solution for remote locations where motorised vehicle transportation options are limited
  • Collapsible when empty for efficient storage and transport, or for efficient packing into small spaces like back packs for remote sampling expeditions
  • Unprocessed container comes assembled and ready for own cleaning procedure
  • With white LDPE-lined PP cap (