Tisk…
Water sampling bottles, HDPE, narrow/wide neck
Dodavatel: EP SCIENTIFIC
313-1000 312-0120 312-0950 313-0125 313-0250 313-0500 112-2000 112-4000 312-4000BPC 112-0500 112-0120 112-0250BPC 112-0500BPC 112-0950 112-0950BPC 112-2000BPC 112-0250 112-4000BPC 312-0250BPC 312-0500 312-0500BPC 312-0950BPC 312-2000 312-4000 312-0250 312-0120BPC
EPSC313-1000EA 3130 CZK
EPSC313-1000 EPSC312-0120 EPSC312-0950 EPSC313-0125 EPSC313-0250 EPSC313-0500 EPSC112-2000 EPSC112-4000 EPSC312-4000BPC EPSC112-0500 EPSC112-0120 EPSC112-0250BPC EPSC112-0500BPC EPSC112-0950 EPSC112-0950BPC EPSC112-2000BPC EPSC112-0250 EPSC112-4000BPC EPSC312-0250BPC EPSC312-0500 EPSC312-0500BPC EPSC312-0950BPC EPSC312-2000 EPSC312-4000 EPSC312-0250 EPSC312-0120BPC
Water sampling bottles, HDPE, narrow/wide neck
Láhve Láhve na odběr vzorků vody
HDPE containers, white, with LDPE-lined polypropylene cap, narrow/wide neck.
  • Narrow mouth for easy handling and precision pouring
  • Wide mouth for easy filling and sample retrieval
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Each container individually barcoded with lot number and unique container number
These containers are recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes. Certified to meet EPA performance basedMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí NAR Certificate of Retirement