Tisk…
Bottles, wide neck, with thread, without screw cap
Dodavatel: EP SCIENTIFIC
012-0120BP 012-0250BP 012-0500BP 012-0950BP
EPSC012-0120BPEA 16120 CZK
EPSC012-0120BP EPSC012-0250BP EPSC012-0500BP EPSC012-0950BP
Bottles, wide neck, with thread, without screw cap
Láhve Láhve plastové
HDPE, translucent.
  • Without screw caps
  • Wide mouth for easy filling and sample retrieval
  • Recommended for general purpose sampling where cleaning and certification is not required
  • Economically bulk packed without caps
Wide-mouth packers are general-purpose containers, bulk and unassembled. Unprocessed, ready for own cleaning procedure and assembly with screw caps purchased separately.