Tisk…
Mastercycler® nexus X2 PCR Thermal Cycler
Dodavatel: EPPENDORF
6336000.040 6337000.043 6336000.015 6338000.012 6339000.016
731-0431EA 259350 CZK
731-0431 EPPE6337000.043 732-3666 EPPE6338000.012 EPPE6339000.016
Mastercycler® nexus X2 PCR Thermal Cycler
PCR cyklery
For a high PCR workload, the Eppendorf® Mastercycler nexus X2 offers efficiency with two blocks (32-well and 64-well), allowing simultaneous runs of two independent protocols. Accommodates a range of consumables up to 0,5 ml tubes. For increased capacity, connect with up to three additional units.
  • Asymmetrically divided block for running different assays in parallel, one 32-well and one 64-well block
  • Intuitive graphic programming
  • Universal block for strips, 0,1, 0,2 and 0,5 ml PCR tubes and fits for divisible PCR plates
  • Small footprint
  • Optional gradient
More Product Information