Tisk…
Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
Dodavatel: EPPENDORF
4925000.030 4925000.049 4925000.014 4925000.022 4925000.057 4925000.065 4925000.073 4925000.081 4925000.090 4925000.103 4925000.111 4925000.120 4925000.138 4925000.146 4925000.154 4925000.162 4925000.170 4924000.010 4924000.029 4924000.037 4924000.045 4924000.053 4924000.061 4924000.070 4924000.088 4924000.096 4924000.100 4924000.118 4924000.908 4924000.916 4924000.924 4924000.940
EPPE4925000.030EA 6100 CZK
EPPE4925000.030 EPPE4925000.049 EPPE4925000.014 EPPE4925000.022 EPPE4925000.057 EPPE4925000.065 EPPE4925000.073 EPPE4925000.081 EPPE4925000.090 EPPE4925000.103 EPPE4925000.111 EPPE4925000.120 EPPE4925000.138 EPPE4925000.146 EPPE4925000.154 EPPE4925000.162 EPPE4925000.170 613-5832 613-5833 613-5834 613-5835 613-5836 613-5837 613-5838 613-5839 613-5840 613-5841 613-5842 EPPE4924000.908 EPPE4924000.916 EPPE4924000.924 613-7086
Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
Pipety Micropipettes
The robust Reference® 2 is the successor to the Reference® pipette and features a new design, reduced weight and unique one-button operation.
  • Single button operation and spring loaded tip cone for fast and ergonomic handling with reduced operating effort
  • Quick tip ejection with active aerosol reduction; haptic feedback prevents accidential tip ejection
  • Serial number is printed on multiple components of the pipette for heightened traceability
  • Embedded RFID chip containing all relevant data ensures complete traceability
  • Optional TrackIT software allows
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Certifikát ochrany životního prostředí Renewable Energy Certificate, ISO 14001 Renewable Energy Certificate, ISO 14001 Renewable Energy Certificate, ISO 14001