Tisk…
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Dodavatel: EPPENDORF
0030108.094 0030108.116 0030108.132 0030108.302 0030122.216 0030122.240 0030122.356 0030108.442 0030108.450 0030108.434 0030122.283
525-0132EA 462 CZK
525-0132 525-0133 525-0134 525-0792 EPPE0030122.216 EPPE0030122.240 EPPE0030122.356 EPPE0030108.442 EPPE0030108.450 EPPE0030108.434 EPPE0030122.283
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Zkumavky Mikrozkumavky centrifugační
Engineered to minimise sample binding and elevate recovery, Eppendorf® Protein LoBind® tubes safeguard valuable proteins during processing. The cutting-edge LoBind® technology ensures optimal yields for Western blotting, mass spectrometry, and protein purification applications.
  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors
  • Available in tube, microplate, and deep-well plate formats for convenient
More Product Information