Tisk…
Recombinant dengue NS1 ST3
Dodavatel: ENZO LIFE SCIENCES
ENZPRT1060100
ENZOENZPRT1060100EA 23890 CZK
ENZOENZPRT1060100
Recombinant dengue NS1 ST3
Proteiny
Dengue virus NS1 protein serotype 3 presented in its native folded state with all post-translational modifications to deliver optimal antigenicity.
Produced in HEK 293 cells. Dengue Viral NS1 Serotype 3 (DENV1-NS1) from strain Sri Lanka D3/H/IMTSSA-SRI/2000/1266.