Tisk…
BNP fragment ELISA kit, grow cells
Dodavatel: ENZO LIFE SCIENCES
ENZKIT1520001
ENZOENZKIT1520001EA 21810 CZK
ENZOENZKIT1520001
BNP fragment ELISA kit, grow cells
Test ELISY
Sensitive BNP fragment ELISA kit enabling detection of BNP fragment in serum and plasma.
  • Quantitative determination of human BNP fragment
  • Colorimetric, competitive enzyme immunoassay
For the quantitative determination of human BNP fragment in serum and plasma. The BNP Fragment ELISA kit is a colorimetric competitive enzyme immunoassay kit.