Tisk…

Anti-CD38 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: HB7]

Dodavatel: ENZO LIFE SCIENCES
ENZABS1590100
ENZOENZABS1590100EA 19870 CZK
ENZOENZABS1590100
Anti-CD38 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: HB7]
Protilátky
Recommended Applications: Flow Cytometry

Species reactivity: Human

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR