Tisk…

Hydroxystilbamidine bis(methanesulphonate) ≥95% (by HPLC), Ultrapure

Dodavatel: ENZO LIFE SCIENCES

ENZ52253
ENZOENZ52253EA 6590 CZK
ENZOENZ52253
Hydroxystilbamidine bis(methanesulphonate) ≥95% (by HPLC), Ultrapure
Hydroxystilbamidine bis(methanesulphonate)

Neuronal detection


Hydroxystilbamidine is a cationic dye, equivalent to Fluoro- Gold™, which is frequently used as a retrograde neuronal tracer. The fluorescent tracer provides intense retrograde labeling that is extremely sensitive and reliable, does not diffuse out of retrograde-labeled neurons, and can be pressure-injected or introduced by iontophoresis into cells. Hydroxystilbamidine is compatible with most frequently used neuroanatomical techniques such as immunofluorescence, immunocytochemistry, autoradiography, and horseradish peroxidase-based histochemistry, paraffin embedding. Wavelength Maxima: Excitation 385 nm, Emission 536 nm.

Vzorec: C₁₈H₂₄N₄O₇S₂
Storage Temperature: Freezer
MDL Number: MFCD01863083
CAS číslo: 223769-64-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR