Tisk…

Di-8-ANEPPS 90 HPLC_ASSAY_METHOD stain_membrane, Histology

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 4-(2-[6-(Dioctylamino)-2-naphthalenyl]ethenyl)-1-(3-sulfopropyl)pyridinium inner salt

ENZ52204
ENZOENZ52204EA 12860 CZK
ENZOENZ52204
Di-8-ANEPPS 90 HPLC_ASSAY_METHOD stain_membrane, Histology
Di-8-ANEPPS

Membrane Potential Detector


Di-8-ANEPPS is a member of the ANEP class of membrane potential dyes. These dyes are weakly fluorescent in aqueous media, and become strongly fluorescent upon binding to lipophilic environments such as membranes. A change in the surrounding electronic field demonstrates a membrane potential-dependent shift in excitation spectra. Di-8-ANEPPS is less susceptible for cellular internalization than other ANEP dyes probably due to a sulfonate group. The response is sufficiently fast to detect transient (millisecond) potential changes in excitable cells, including single neurons, cardiac cells and intact brains. Wavelength Maxima: Excitation 498nm, Emission 713nm

Vzorec: C₃₆H₅₂N₂O₃S
MW: 592.87 g/mol
MDL Number: MFCD03453608
CAS číslo: 157134-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8320b5cf6026376fdc4e3f6e7c56743b93e74882&vid=b4fda880de1f12fdcf1c9c9e683f7641967a4fa2&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR