Tisk…
([K11-only]Ub)n-ubiquitinylated Substrate
Dodavatel: ENZO LIFE SCIENCES
BMLUW06200025
ENZOBMLUW06200025EA 8640 CZK
ENZOBMLUW06200025
([K11-only]Ub)n-ubiquitinylated Substrate
Proteiny
Protein components produced in E. coli. Synthesized enzymatically in vitro. (K11-only)Ubn-ubiquitinylated substrate protein was produced using K11-only mutant ubiquitin.