Tisk…
Human recombinant MMP-19 (catalytic domain)
Dodavatel: ENZO LIFE SCIENCES
BMLSE5610010
ENZOBMLSE5610010EA 10640 CZK
ENZOBMLSE5610010
Human recombinant MMP-19 (catalytic domain)
Proteiny
Produced in E.coli.
Active recombinant MMP-19. MMP-19 is expressed widely, and degrades aggrecan, fibronectin, type I gelatin, and basement membrane components such as laminin, nidogen, and type IV collagen. It is involved in angiogenesis, cancer, rheumatoid arthritis, multiple sclerosis, and psoriasis.