Tisk…

Gabexate mesylate ≥98% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

BMLPI1440010 BMLPI1440050
ENZOBMLPI1440010EA 2540 CZK
ENZOBMLPI1440010 ENZOBMLPI1440050
Gabexate mesylate ≥98% (by HPLC)
Gabexate mesylate

A competitive, reversible inhibitor of serine proteases tryptase (Ki=3.4nM), thrombin (Ki=500nM), uPA (Ki=1.3µM), plasmin (Ki=1.5µM), trypsin (Ki=2µM), factor Xa (Ki=2µM). At higher concentrations it inhibits kallikrein-1 (Ki=2.6mM), C1 esterase (IC50=520µM), as well as phospholipase A2 (IC50=500µM), rat iNOS (Ki=5mM) and cNOS (Ki=150µM), and swine kidney copper amine oxidase (Ki=27µM), but not chymotrypsin. Has been used in tissue culture and in vivo. In clinical use for pancreatitis.


  • Protease inhibitor


White solid.


Soluble in water (25 mg/ml).

Vzorec: C₁₆H₂₃N₃O₄·CH₃SO₃H
Storage Temperature: Freezer
MDL Number: MFCD00210299
CAS číslo: 56974-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00639d19f24fb21bb7818076610e72d9744d8e68&vid=fe6afd23d45db691f54b97b16619e333dec1beb3&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR