Tisk…

6-Ketoprostaglandin-F1α 98 TLC_ASSAY_METHOD

Dodavatel: ENZO LIFE SCIENCES
Varování

Synonyms: 6-Keto-PGF1α

BMLPG0170001
ENZOBMLPG0170001EA 4080 CZK
ENZOBMLPG0170001
6-Ketoprostaglandin-F1α 98 TLC_ASSAY_METHOD
6-Ketoprostaglandin-F1alpha

Biosynthesis via hydrolysis of prostacyclin. A reliable parameter for measurement of prostacyclin. Ultra-pure.


Soluble in ethanol (25 mg/ml).

Vzorec: C₂₀H₃₄O₆
MW: 370.486 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135222
CAS číslo: 58962-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a72862d64466bc623b427885fb0d72c7807fd82f&vid=85a06c7f71f88afdf3303c2b41bbfd7601a6c0bb&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Purity ≥98% (TLC)
Appearance White to off-white solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR