Tisk…

Prostaglandin F2α tris salt (synthetic) ≥99% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

BMLPG0090010
ENZOBMLPG0090010EA 9980 CZK
ENZOBMLPG0090010
Prostaglandin F2α tris salt (synthetic) ≥99% (by HPLC)
Prostaglandin F2alpha tris salt

A major uterine luteolytic prostaglandin. Endogenous agonist of the prostanoid FP receptor (Kd=1.0 nM). Inhibits differentiation of 3T3-L1 preadipocytes. The product is not sterile.


  • Prostanoid receptor ligand


White solid.


Soluble in water.

Vzorec: C₂₄H₄₅NO₈
Storage Temperature: Freezer
MDL Number: MFCD00077863
CAS číslo: 38562-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73ad96bf39433ed084ed3fa3ae57c6e50c76387b&vid=57bc6e65095a18752f8ad4fd274aad935d9c7c98&prid=web&clid=SAPDAM

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR