Tisk…

SCH-202676 ≥99% (by HPLC), yellow solid

Dodavatel: ENZO LIFE SCIENCES
Varování

BMLG5210010
ENZOBMLG5210010EA 4590 CZK
ENZOBMLG5210010
SCH-202676 ≥99% (by HPLC), yellow solid
SCH-202676

Blocks agonist and antagonist binding to G protein-coupled receptors.


SCH-202676


Soluble in DMSO (7 mg/ml, with warming).

Vzorec: C₁₅H₁₃N₃S·HBr
MW: 348.3 g/mol
Storage Temperature: Ambient
CAS číslo: 70375-43-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fbcd00e5afae87a2439715e67dfcfa393b6b350&vid=7301b8ced0780bf681a29c1135aac0d5633cb07b&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Purity ≥99% (HPLC)
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR