Tisk…

Indirubin-3'-oxime ≥98%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 3-(Hydroxyimino)-[2,3'-biindolinylidene]-2'-one, Indirubin-3'-monoxime

BMLCC2070005
ENZOBMLCC2070005EA 12600 CZK
ENZOBMLCC2070005
Indirubin-3'-oxime ≥98%
Indirubin-3'-oxime

A potent inhibitor of glycogen synthase kinase-3β (IC50=22 nM). Also inhibits CDK1 (p34cdc2; IC50=180 nM) and CDK5 (IC50=100 nM). It reversibly arrests asynchronous HBL-100 cells at G2. Induces apoptosis in the mammary carcinoma cell line MCF-7 (10 µM).


  • Inhibitor of CDK and GSK


Dark red solid.


Soluble in DMSO (6mg/ml) and ethanol (4mg/ml).

Vzorec: C₁₆H₁₁N₃O₂
MW: 277.283 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02683594
CAS číslo: 160807-49-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR