Tisk…

Noc-7 ≥96% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES
Varování

ALX430018M005 ALX430018M010
ENZOALX430018M005EA 3510 CZK
ENZOALX430018M005 ENZOALX430018M010
Noc-7 ≥96% (by HPLC)
Noc-7

Nitric oxide (NO) donor.


The half-life time of 0.1 mM NOC-7 is 5 min at 37 °C in 0.1 M PBS, pH 7.4.


Reconstitute with 0.1 M sodium hydroxide solution to 10 - 50 mM.


Upozornení: Protect from light and moisture. 

Vzorec: C₅H₁₄N₄O₂
Storage Temperature: Freezer
CAS číslo: 146724-84-7

Objednat

Výsledky specifikačního testu

Purity ≥96% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR