Tisk…

Pterostilbene ≥98% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

ALX385034M100
ENZOALX385034M100EA 7330 CZK
ENZOALX385034M100
Pterostilbene ≥98% (by HPLC)
Pterostilbene

CYP1A1 inhibitor


Cell permeable natural methoxylated analog of resveratrol (Prod. No. BML-FR104). Antioxidant, antiproliferative, anti-inflammatory, anti-hyperglycemic and anti-diabetic agent. Induces apoptosis. Induces Autophagy. Very potent inhibitor of cytochrome P450 1A1 (CYP1A1) with Ki=0.57µM.Inhibits activator protein 1 (AP-1) and NF-kappaB activation. Moderately inhibits COX-1 and COX-2 (IC50=19.8µM and 83.9µM).


Synthetic.

Vzorec: C₁₆H₁₆O₃
MDL Number: MFCD00238710
CAS číslo: 537-42-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR