Tisk…

Rottlerin ≥98% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: (E)-1-(6-(3-Acetyl-2,4,6-trihydroxy-5-methylbenzyl)-5,7-dihydroxy-2,2-dimethyl-2H-chromen-8-yl)-3-phenylprop-2-en-1-one

ALX350075M010 ALX350075M025
ENZOALX350075M010EA 3000 CZK
ENZOALX350075M010 ENZOALX350075M025
Rottlerin ≥98% (by HPLC)
Rottlerin

PKCdelta inhibitor


Mitochondrial uncoupler that depolarizes the mitochondrial membrane potential, reduces cellular ATP levels, activates 5‘-AMP-activated protein kinase (AMPK) and affects mitochondrial production of reactive oxygen species (ROS). Potent activator of multiple Ca²⁺-sensitive K+ channels. Blocks several kinases and non-kinase proteins in vitro. Has been widely-used as a selective inhibitor of protein kinase Cdelta (PKCdelta) (IC50=3-6µM). Although there is extensive published documentation to support the use of rottlerin as a selective PKCdelta inhibitor, there has been some controversy in the literature over this claim. Some of this controversy may arise from the fact that rottlerin is a promiscuous inhibitor and therefore may not always display the same properties as would be expected from a classical 1:1 inhibitor. However, there are studies indicate that rottlerin has no direct effect on PKCdelta and that it should not be used to determine the involvement of PKCdelta in biological processes. Induces autophagy by inhibition of mTORC1 signaling.


Isolated from Mallotus philippinensis.

Vzorec: C₃₀H₂₈O₈
MDL Number: MFCD00017361
CAS číslo: 82-08-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR