Tisk…

Genistein

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

ALX350006G001 ALX350006M010 ALX350006M025 ALX350006M050 ALX350006M100
ENZOALX350006G001EA 28790 CZK
ENZOALX350006G001 ENZOALX350006M010 ENZOALX350006M025 ENZOALX350006M050 ENZOALX350006M100
Genistein
Genistein
Vzorec: C₁₅H₁₀O₅
MW: 270.24
Bod tání: 297…298 °C
MDL Number: MFCD00016952
CAS číslo: 446-72-0
EINECS: 207-174-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR