Tisk…

SB 225002 ≥98% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

ALX270478M001 ALX270478M005
ENZOALX270478M001EA 1730 CZK
ENZOALX270478M001 ENZOALX270478M005
SB 225002 ≥98% (by HPLC)
SB 225002

Chemokine receptor ligand


Potent and selective antagonist of CXCR2 chemokine receptor (IC50=22nM). >150-fold more selective over CXCR1 receptor. Inhibits IL-8- (IC50=8nM) and GROα-mediated (IC50=10nM) calcium mobilisation. Prevents IL-8-induced neutrophil migration. Inhibits HIV replication in lymphocytes and macrophages.

Vzorec: C₁₃H₁₀BrN₃O₄
MDL Number: MFCD00954637
CAS číslo: 182498-32-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR