Tisk…

N-Oxalylglycine ≥98%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 2-((Carboxymethyl)amino)-2-oxoacetic acid

ALX270412M010
ENZOALX270412M010EA 1920 CZK
ENZOALX270412M010
N-Oxalylglycine ≥98%
N-Oxalylglycine

PHD inhibitor


Competitive inhibitor of PHDs (prolyl hydroxylase domain-containing proteins; HIF-PHs).

Vzorec: C₄H₅NO₅
MW: 147.09 g/mol
MDL Number: MFCD00913253
CAS číslo: 5262-39-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR