Tisk…

PJ-34 hydrochloride synthetic 98 HPLC_ASSAY_METHOD

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 2-(Dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride, PJ34 HCl

ALX270289M001 ALX270289M005 ALX270289M025
ENZOALX270289M001EA 2390 CZK
ENZOALX270289M001 ENZOALX270289M005 ENZOALX270289M025
PJ-34 hydrochloride synthetic 98 HPLC_ASSAY_METHOD
PJ-34 hydrochloride

Potent, water soluble poly(ADP-ribose) polymerase (PARP) inhibitor (EC50=20nM compared to EC50=200µM of the prototypical PARP inhibitor 3-aminobenzamide). Inhibits peroxynitrite-induced cell necrosis (EC50=20nM). Has significant, dose-dependent, anti-inflammatory effects in a variety of local inflammation models and provides cardioprotection by decreasing myocardial infarct size.


  • PARP inhibitor


White to light brown or yellow powder.


Soluble in DMSO (up to 30 mg/ml) or water (up to 20 mg/ml).

Vzorec: C₁₇H₁₇N₃O₂·HCl
MW: 295.2·36.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22581510
CAS číslo: 344458-15-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR