Tisk…

Tyrphostin AG 1024 ≥98% (by HPLC and UV method)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 2-(3-Bromo-5-(tert-butyl)-4-hydroxybenzylidene)malononitrile, AG1024

ALX270217M001 ALX270217M005
ENZOALX270217M001EA 3400 CZK
ENZOALX270217M001 ENZOALX270217M005
Tyrphostin AG 1024 ≥98% (by HPLC and UV method)
Tyrphostin AG 1024

Specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor kinases. Induces apoptosis by downregulation of the expression of phospho-Akt1 and Bcl-2. Also increases Bax, p53 and p21 expression.


  • IGF-1 inhibitor


Yellow powder.


Soluble in acetonitrile (5mg/ml), DMSO (8mg/ml), dichloromethane (23mg/ml) or methanol (5mg/ml); insoluble in 100% ethanol.

Vzorec: C₁₄H₁₃BrN₂O
Storage Temperature: Freezer
MDL Number: MFCD02179365
CAS číslo: 65678-07-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR