Tisk…

Piceatannol

Dodavatel: ENZO LIFE SCIENCES

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

ALX270202M001 ALX270202M010 ALX270202M050
ENZOALX270202M001EA 1670 CZK
ENZOALX270202M001 ENZOALX270202M010 ENZOALX270202M050
Piceatannol
Piceatannol
Vzorec: C₁₄H₁₂O₄
MDL Number: MFCD00221715
CAS číslo: 10083-24-6
New Product

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR