Tisk…

Bisindolylmaleimide III ≥98%

Dodavatel: ENZO LIFE SCIENCES

ALX270051M001 ALX270051M005
ENZOALX270051M001EA 2170 CZK
ENZOALX270051M001 ENZOALX270051M005
Bisindolylmaleimide III ≥98%
Bisindolylmaleimide III

PKC inhibitor


Potent and selective inhibitor of protein kinase C (PKC).

Vzorec: C₂₃H₂₀N₄O₂
MDL Number: MFCD01076633
CAS číslo: 137592-43-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR