Tisk…
Mca-Pro-Leu-Ala-Nva-Dap(Dnp)-Ala-Arg-NH2
Dodavatel: ENZO LIFE SCIENCES
ALX260123M001
ENZOALX260123M001EA 10520 CZK
ENZOALX260123M001
Mca-Pro-Leu-Ala-Nva-Dap(Dnp)-Ala-Arg-NH2
Proteiny
MMP-2 substrate
Fluorogenic substrate for MMP-2.