Tisk…

Anti-CXCR4 Goat Polyclonal Antibody

Dodavatel: ENZO LIFE SCIENCES
ALX210819C200
ENZOALX210819C200EA 15910 CZK
ENZOALX210819C200
Anti-CXCR4 Goat Polyclonal Antibody
Protilátky
Host: Goat

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes mouse CXCR4.

Type: Primary
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR