Tisk…

L-Thiocitrulline dihydrochloride ≥97%

Dodavatel: ENZO LIFE SCIENCES

ALX106013M010 ALX106013M050
ENZOALX106013M010EA 2880 CZK
ENZOALX106013M010 ENZOALX106013M050
L-Thiocitrulline dihydrochloride ≥97%
L-Thiocitrulline dihydrochloride

NOS inhibitor


Inhibitor of neuronal (nNOS; NOS I; Ki=60nM) and inducible (iNOS; NOS II) nitric oxide synthase.


Upozornení: Research use only.

Storage Temperature: Ambient
CAS číslo: 212051-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c47704dc56ba9c94ce075e09668bf90bb879b62&vid=044aaea4a2963043961ef6b53e91d57197782260&prid=web&clid=SAPDAM
New Product

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR