Tisk…
Anti-ERP72 Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADISPS720D ADISPS720F
ENZOADISPS720DEA 6870 CZK
ENZOADISPS720D ENZOADISPS720F
Anti-ERP72 Rabbit Polyclonal Antibody
Protilátky
Endoplasmic reticulum protein (ERp) 72, a 72 kDa protein localized to the endoplasmic reticulum, is a member of the Protein Disulfide Isomerase (PDI) family of proteins. Together with other ER resident proteins such as BiP, GRP94, and PDI, they serve as the molecular chaperones for proper folding of newly translocated and glycolsylated proteins such as Thyroglobulin (Tg) and Human chorionic gonadotropin (hCG)-beta.
  • For Immunofluorescence, Immunohistochemistry, Western Blot
Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey andMore Product Information