Tisk…
Anti-HSP90B Mouse Monoclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADISPA842D ADISPA842F
ENZOADISPA842DEA 8270 CZK
ENZOADISPA842D ENZOADISPA842F
Anti-HSP90B Mouse Monoclonal Antibody
Protilátky
The Hsp90 family of heat shock proteins represents one of the most abundantly expressed and highly conserved families of cellular chaperones whose expression can be upregulated under conditions of cellular stress, and includes cytoplasmic (Hsp90-alpha/beta), ER (grp94), and mitochondrial (TRAP1) localized members. Structurally, Hsp90 is characterized by an N-terminal ATP-binding domain, a medial substrate-binding domain, and a C-terminal dimerization motif. Hsp90 dimers function in cooperation with cochaperones (e.g. Hsp40, Hsp70, Hop, p23) to stabilize a multitude of client proteinMore Product Information