Tisk…
Anti-BAD Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADIAAP020D ADIAAP020F
ENZOADIAAP020DEA 6990 CZK
ENZOADIAAP020D ENZOADIAAP020F
Anti-BAD Rabbit Polyclonal Antibody
Protilátky
Bad is an important regulatory protein involved in the control of cell survival and apoptosis, and is a member of the BH3-only family of proteins such as Bim and Bid that share homology with Bcl-2 only in the BH3 domain. Phosphorylated Bad is sequestered in the cytosol by 14-3-3 proteins under normal conditions, but is dephosphorylated and activated following apoptotic signals. Apoptotic stimuli induce Bad dephosphorylation by a number of phosphatases (e.g., calcineurin) and recruitment to the mitochondria, where Bad heterodimerizes with and inhibits prosurvival proteins of the Bcl family.More Product Information
New Product
Objednat
Produktová specifikace
Symbol antigenu BAD BAD
Název antigenu BCL2-associated agonist of cell death BCL2-associated agonist of cell death
Environmentally Preferable
Synonyma antigenu BCL2L8|Bcl-2-binding component 6|BBC6|Bcl2 antagonist of cell death|BBC2|BAD BCL2L8|Bcl-2-binding component 6|BBC6|Bcl2 antagonist of cell death|BBC2|BAD
Druh protilátky Primární Primární
Klonovalita Polyclonal Polyclonal
Reaktivita Mouse
Hostitel Rabbit Rabbit
Izotyp
Western blot Yes Yes
Imunochemie Yes Yes
Formulář Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Čištění Protein A affinity purified