Tisk…
IHC mount, HIGHDEF®
Dodavatel: ENZO LIFE SCIENCES
ADI9502610030
ENZOADI9502610030EA 2610 CZK
ENZOADI9502610030
IHC mount, HIGHDEF®
Barviva Microscopy and staining reagents
IHC mount is an aqueous-based permanent, high stability mounting medium designed for use with chromogenic labeling. Use for mounting of tissue specimens stained with peroxidase and alkaline phosphatase-based systems and various fluorescent dyes.
  • Flexible formulation optimised for AEC, DAB, Fast Red, BCIP/NBT, and BCIP/INT
  • High definition visualisation for IHC, IF, ISH, FISH
  • Allows permanent storage of tissue specimens without fading of the chromogens
  • No coverslip required
  • No exposure to organic fumes
Formulation: Liquid, mounting medium containing <0,1% sodium azide.