Tisk…
IHC diluent (primary Ab)
Dodavatel: ENZO LIFE SCIENCES
ADI9502440250
ENZOADI9502440250EA 4540 CZK
ENZOADI9502440250
IHC diluent (primary Ab)
Test Imunologická stanovení
Immunohistochemistry diluent (primary Ab) can be used as a diluent to prepare primary antibodies and negative controls.
  • Optimal pH with stabilising proteins in the IHC diluent (primary Ab)
  • Maximum stability of primary antibodies
  • Good penetration of primary antibodies via selected concentrations of sodium chloride and detergents
Specifically designed to stabilise primary antibodies and reduce non-specific background staining observed during immunostaining.