Tisk…
SLPI (human), ELISA kit
Dodavatel: ENZO LIFE SCIENCES
ADI9002220001
ENZOADI9002220001EA 25460 CZK
ENZOADI9002220001
SLPI (human), ELISA kit
Test ELISY
Rapid ELISA kit to quantify SLPI, a secretory leukocyte peptidase inhibitor and unique cancer biomarker.
For the quantitative determination of human SLPI (ALP, Antileukoproteinase) in serum.
The SLPI ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of SLPI in serum with results in just 2.5 hours.
Informace pro objednávání: Contains Microtiter Plate, Standards, Calibrators, Antibody-conjugate, TMB Reagent, Stop Solution, Wash Buffer Concentrate.