Tisk…
SOD activity kit
Dodavatel: ENZO LIFE SCIENCES
ADI900157
ENZOADI900157EA 24430 CZK
ENZOADI900157
SOD activity kit
Test Buněčná stanovení
ELS offers a large collection of superoxide dismutase products for oxidative stress research.
For the measurement of total SOD activity in cell lysates and tissue from any species.
This is a versatile assay for the determination of SOD activity. Differentiate between the activity of three SOD isoenzymes: MnSOD, FeSOD, and Cu/ZnSOD. Obtain higher throughput sample analysis with a convenient colorimetric 96 well plate format and fast results in just 10 minutes.
Informace pro objednávání: Contents: Microtiter plate, Standard, SOD buffer concentrate, Xanthine oxidase, Xanthine solution, Triton x-100, WST-1 reagent