Tisk…
6-keto-PGF1α ELISA Kits
Dodavatel: ENZO LIFE SCIENCES
ADI900004
ENZOADI900004EA 14480 CZK
ENZOADI900004
6-keto-PGF1α ELISA Kits
Test ELISY
Highly sensitive ELISA, detecting as low as 1.40pg/ml in just 3 hours.
For the quantitative determination of 6-keto-PGF1α in culture supernatants, serum, saliva, and urine from any species.
The 6-keto-PGF1α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 1α EIA kit has been fully validated in-house in a wide variety of sample matrices.
Informace pro objednávání: Contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-272 (5x96 wells).